Anti-PPP1R1A Rabbit Polyclonal Antibody
Catalog # AVIVARP42314_P050
Supplier: Aviva Systems Biology
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:protein phosphatase 1, regulatory (inhibitor) subunit 1A
- Antigensymbol:PPP1R1A
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Western blot:Yes
- Epitop:N-terminal
- Korsad adsorption:none
- Gen-ID:58200
- Antigen synonymer:I-1|0610038N18Rik
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:50 µG
Specifications
About this item
Type: Primary
Antigen: PPP1R1A (protein phosphatase 1, regulatory (inhibitor) subunit 1A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: