Anti-MYH1 Rabbit Polyclonal Antibody
AVIVARP42269_P050
- Antikroppstyp:Primär
- Antikroppsnamn:myosin, heavy chain 1, skeletal muscle, adult
- Antigensymbol:MYH1
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Western blot:Yes
- Epitop:N-terminal
- Korsad adsorption:none
- Gen-ID:4619
- Antigen synonymer:MYHa|HEL71|MYHSA1|MyHC-2x|MyHC-2X/D
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:50 µG
Type: Primary
Antigen: MYH1 (myosin, heavy chain 1, skeletal muscle, adult)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: