Anti-MAK Rabbit Polyclonal Antibody
AVIVARP42268_P050
- Antikroppstyp:Primär
- Antikroppsnamn:male germ cell-associated kinase
- Antigensymbol:MAK
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Western blot:Yes
- Epitop:C-terminal
- Korsad adsorption:none
- Gen-ID:4117
- Antigen synonymer:RP62|dJ417M14.2
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:50 µG
Type: Primary
Antigen: MAK (male germ cell-associated kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: