Anti-DHRS2 Rabbit Polyclonal Antibody
Katalog # AVIVARP42259_P050
Leverantör: Aviva Systems Biology
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:dehydrogenase/reductase (SDR family) member 2
- Antigensymbol:DHRS2
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Western blot:Yes
- Epitop:middle
- Korsad adsorption:none
- Gen-ID:10202
- Antigen synonymer:SDR25C1|HEP27
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:50 µG
Specifikationer
Om denna produkt
Type: Primary
Antigen: DHRS2 (dehydrogenase/reductase (SDR family) member 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: