Anti-EBI3 Rabbit Polyclonal Antibody
AVIVARP42257_P050
- Antikroppstyp:Primär
- Antikroppsnamn:Epstein-Barr virus induced 3
- Antigensymbol:EBI3
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Reaktivitet:Human
- Western blot:Yes
- Epitop:middle
- Korsad adsorption:none
- Gen-ID:10148
- Antigen synonymer:IL-27B|IL27B
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:50 µG
Type: Primary
Antigen: EBI3 (Epstein-Barr virus induced 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human