Anti-CYP2D6 Rabbit Polyclonal Antibody
Katalog # AVIVARP41676_P050
Leverantör: Aviva Systems Biology
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:cytochrome P450, family 2, subfamily D, polypeptide 6
- Antigensymbol:CYP2D6
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Western blot:Yes
- Epitop:middle
- Korsad adsorption:none
- Gen-ID:1565
- Antigen synonymer:P450C2D|CYP2D7BP|P450-DB1|CYP2DL1|P450DB1|CYP2D7AP|CYP2D8P2|CYP2D7P2|CYPIID6|CYP2D|CPD6
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:50 µG
Specifikationer
Om denna produkt
Type: Primary
Antigen: CYP2D6 (cytochrome P450, family 2, subfamily D, polypeptide 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: