Anti-PCBP4 Rabbit Polyclonal Antibody
Katalog # AVIVARP40944_P050
Leverantör: Aviva Systems Biology
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:Poly(rC) binding protein 4
- Antigensymbol:PCBP4
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Western blot:Yes
- Epitop:C-terminal
- Korsad adsorption:none
- Gen-ID:57060
- Antigen synonymer:LIP4|CBP|MCG10
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:50 µG
Specifikationer
Om denna produkt
Type: Primary
Antigen: PCBP4 (poly(rC) binding protein 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: