Anti-DAZAP1 Rabbit Polyclonal Antibody
Katalog # AVIVARP40927_T100
Leverantör: Aviva Systems Biology
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:DAZ associated protein 1
- Antigensymbol:DAZAP1
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Western blot:Yes
- Epitop:C-terminal
- Korsad adsorption:none
- Gen-ID:26528
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:100 µG
Specifikationer
Om denna produkt
Type: Primary
Antigen: DAZAP1 (DAZ associated protein 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: