Anti-NOP56 Rabbit Polyclonal Antibody
Katalog # AVIVARP40644_P050
Leverantör: Aviva Systems Biology
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:NOP56 ribonucleoprotein
- Antigensymbol:NOP56
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Reaktivitet:Human
- Western blot:Yes
- Epitop:middle
- Korsad adsorption:none
- Gen-ID:10528
- Antigen synonymer:SCA36|NOL5A
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:50 µG
Specifikationer
Om denna produkt
Type: Primary
Antigen: NOP56 (NOP56 ribonucleoprotein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human