Anti-SRSF6 Rabbit Polyclonal Antibody
Katalog # AVIVARP40632_P050
Leverantör: Aviva Systems Biology
Specifikationer
- Antikroppstyp:Primär
- Antikroppsnamn:Serine/arginine-rich Splicing Factor 6
- Antigensymbol:SRSF6
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Reaktivitet:Zebrafish
- Western blot:Yes
- Epitop:middle
- Korsad adsorption:none
- Gen-ID:6431
- Antigen synonymer:SFRS6|SRP55|HEL-S-91|B52
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:50 µG
Specifikationer
Om denna produkt
Type: Primary
Antigen: SRSF6 (serine/arginine-rich splicing factor 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Zebrafish