Anti-PPIE Rabbit Polyclonal Antibody
Katalog # AVIVARP40627_T100
Supplier: Aviva Systems Biology
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:peptidylprolyl isomerase E (cyclophilin E)
- Antigensymbol:PPIE
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Western blot:Yes
- Epitop:middle
- Korsad adsorption:none
- Gen-ID:10450
- Antigen synonymer:CYP-33|CYP33
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:100 µG
Specifications
About this item
Type: Primary
Antigen: PPIE (peptidylprolyl isomerase E (cyclophilin E))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: