Anti-ZNF707 Rabbit Polyclonal Antibody
AVIVARP40004_P050
- Antikroppstyp:Primär
- Antikroppsnamn:zinc finger protein 707
- Antigensymbol:ZNF707
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Western blot:Yes
- Epitop:N-terminal
- Korsad adsorption:none
- Gen-ID:286075
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:50 µG
Type: Primary
Antigen: ZNF707 (zinc finger protein 707)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: