Anti-ZKSCAN5 Rabbit Polyclonal Antibody
Catalog # AVIVARP31830_T100
Supplier: Aviva Systems Biology
Specifications
- Antikroppstyp:Primär
- Antikroppsnamn:zinc finger with KRAB and SCAN domains 5
- Antigensymbol:ZKSCAN5
- Klonalitet:Polyclonal
- Konjugation:Unconjugated
- Värd:Rabbit
- Reaktivitet:Human,Mouse
- Western blot:Yes
- Epitop:N-terminal
- Korsad adsorption:none
- Gen-ID:23660
- Antigen synonymer:ZNF914|ZFP95|ZSCAN37
- Förvaringstemperatur:Cool
- Shipping temperature:Ice
- Förp.:100 µG
Specifications
About this item
Type: Primary
Antigen: ZKSCAN5 (zinc finger with KRAB and SCAN domains 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse