Orders LinkContactUsLinkComponent
Multiple adapters, Quickfit®
Multiple adapters, Quickfit®
24/29 NS, 19/26 NS
Katalog # 201-3002
Leverantör:   SciLabware
Multiple adapters, Quickfit®
Katalog # 201-3002
Leverantör:  SciLabware
Multiple adapters, Quickfit®
Multiple adapters, Quickfit®
24/29 NS, 19/26 NS
Price
1 275,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Produktdetaljer och dokument
PYREX®, borosilicate glass
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With three necks. Two vertical and one angled at 45°.
Specifikationer
  • Kärna:
    24/29 NS
  • NS-hylsa:
    19/26 NS