ContactUsLinkComponent
Dispensing head, Sentry®
Dispensing head, Sentry®
Catalog # ENTEDSU-00-01-00
Supplier:  ENTEGRIS
undefined
Dispensing head, Sentry®
Catalog # ENTEDSU-00-01-00
Supplier:  ENTEGRIS
Specifications
  • Beskrivning:
    Dispense head, single use, Sentry®, standard (corrugated and shroud), without tubing and with FEP encapsulated silicone O ring
  • Förp.:
    1
Specifications
About this item
Designed for use with FluoroPure® single-use HDPE blowmolded drums, this single-use dispense head provides dispensing functions in a single connection.
  • Reliable, high purity o-ring-free dispense connection without chemical key coding
  • All PFA wetted materials provide corrosion resistance and high purity
  • Fill and dispense port comes standard with a 1" Flaretek® flared connection