Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Rör
Mikrorör
Microcentrifuge tubes
Microcentrifuge tubes
Microcentrifuge tubes
Supplier:
Brand
Certificates
Expand image
Microcentrifuge tubes
Supplier:
Brand
Certificates
About this item
PP, translucent.
With lid locking
Frosted marking area
Subdivisions for approx. volume determination
Free of endotoxins, DNase and Rnase
Related Information
Documentation
Certificates
Search by Lot Number
CoC_BRND780540_01W^5127877
CoC_BRND780540_5127877
Documentation