Comfort cartridge set, arium®
Catalog # VIVAH2O-C-PACK
Supplier: Sartorius
Specifications
- Beskrivning:Cartridge set for arium® comfort I and comfort II
- Förp.:1
Specifications
About this item
This is the deionization cartridge set with top-down technology which is optimized for the removal of both organic and inorganic constituents. It is specifically designed to match the unit and delivers ultrapure water which exceeds the ASTM Type 1 quality standard.
- Long lasting high performance and low maintenance operation
- Fast and effective adsorption of impurities through high-grade activated carbon
- Optimised crossflow behavior, prevents separation of the resin mixed-bed
- Patented connection process simplifies the replacement of consumables
Housing is made up of high-grade PP with stainless steel mounting screws. The cleaning medium like Spherical catalytic effective activated carbon ultrapure, semiconductor grade mixed bed ion exchange resin.