Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 kr
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Sweden
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Infärgningsmedel
Microscopy and staining reagents
Immersion oil
Immersion oil
Immersion oil
Catalog # MOTI1101001300011
Supplier:
MOTIC
CAS Number:
SDS
Expand image
Immersion oil
Catalog # MOTI1101001300011
Supplier:
MOTIC
CAS Number:
SDS
Specifications
Förp.:
1
Beskrivning:
Immersion oil 5 ml
Seq:
901
Specifications
About this item
Product Details
Documentation
Product Details
Documentation