Brilliant green bile broth, tubes
Catalog # HACH32215
Supplier: Hach
CAS Number:
Specifications
- Förp.:15
- Standard:-
- Beskrivning:Brilliant green broth tubes, glass
- Volym:15 ml
Specifications
About this item
They are used in the applications of drinking water, wastewater, ambient water, food and beverage. And it is a complete solution for testing drinking water also.
- Saves time
- Saves labor cost for ready to use MPN tubes