Glycine reagent
Katalog # HACH2762133
Leverantör: Hach
CAS-nummer:
Specifikationer
- Kemisk metod:DPD/glycine
- Förp.:29 mL
- Beskrivning:Glycine reagent, 10%, in self contained dropping bottle
Specifikationer
Om denna produkt
Glycine reagent for Hach chlorine dioxide determination by DPD/glycine methods.