ContactUsLinkComponent
Glycine reagent
Glycine reagent
Katalog # HACH2762133
Leverantör:  Hach
CAS-nummer:  
undefined
Glycine reagent
Katalog # HACH2762133
Leverantör:  Hach
CAS-nummer:  
Specifikationer
  • Kemisk metod:
    DPD/glycine
  • Förp.:
    29 mL
  • Beskrivning:
    Glycine reagent, 10%, in self contained dropping bottle
Specifikationer
Om denna produkt
Glycine reagent for Hach chlorine dioxide determination by DPD/glycine methods.