ContactUsLinkComponent
VWR®, Spare Stopcocks
VWR®, Spare Stopcocks
Katalog # 467-0111
Leverantör:  VWR Collection
VWR®, Spare Stopcocks
Katalog # 467-0111
Leverantör:  VWR Collection
Specifikationer
  • Beskrivning:
    Stopcock with PTFE spindle, NS 24/29
  • Seq:
    070
  • Förp.:
    1