ContactUsLinkComponent
Mesh woven filters, PETEX®
Mesh woven filters, PETEX®
Catalog # SEFA07-80/32
Supplier:  SEFAR
Mesh woven filters, PETEX®
Catalog # SEFA07-80/32
Supplier:  SEFAR
Specifications
  • Porstorlek:
    80, 00 µm
  • Seq:
    06
  • Förp.:
    1
Specifications
About this item
Precision woven synthetic monofilament fabrics. It’s a highly specialised monofilament fabric, characterised by precisely defined and controlled, consistent and repeatable material properties such as pore size, thickness, tensile strength, dimensional stability, cleanliness etc.
  • Surface properties are even and consistent
  • Thickness tolerance is within microns through entire rolls and lots
  • Cleanliness meets strict standards