Mesh woven filters, PETEX®
Catalog # SEFA07-80/32
Supplier: SEFAR
Specifications
- Porstorlek:80, 00 µm
- Seq:06
- Förp.:1
Specifications
About this item
Precision woven synthetic monofilament fabrics. It’s a highly specialised monofilament fabric, characterised by precisely defined and controlled, consistent and repeatable material properties such as pore size, thickness, tensile strength, dimensional stability, cleanliness etc.
- Surface properties are even and consistent
- Thickness tolerance is within microns through entire rolls and lots
- Cleanliness meets strict standards