Latex gloves, TouchNTuff® 69-318
Leverantör: Ansell
Certifikat
Om denna produkt
These ambidextrous gloves are resistant to splashes of acids, bases, alcohols and dilute aqueous solutions of most chemicals.
- Flexible and comfortable to wear
- Cooler than vinyl, with superior finger flexibility and grip
- Available in both powdered and powder-free versions
- Powdered gloves have a smooth surface, powder-free gloves have a textured finish
Ideal for use in laboratory analysis, electronic manufacturing and in pharmaceutical industries.
EN 374, Cat. III; AQL 1,5 Food contact approved
Certifications: EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS; Cat. III; AQL 1,5
Food contact approved
Food contact approved
Specifikationer
- Catalog No:
- 112-4265
- 112-4264
- 112-4262
- 112-4266
- 112-4263
- Modell:
- 69-318
- 69-318
- 69-318
- 69-318
- 69-318
- Ej handspecifika/handspecifika:
- Ej handspecifika
- Ej handspecifika
- Ej handspecifika
- Ej handspecifika
- Ej handspecifika
- Engångs/Återanvändbar:
- Engångs
- Engångs
- Engångs
- Engångs
- Engångs
- Pudrad/puderfri:
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Puderfria
- Yta:
- Textured
- Textured
- Textured
- Textured
- Textured
- Kragmodell:
- Med rullad kant
- Med rullad kant
- Med rullad kant
- Med rullad kant
- Med rullad kant
- EN standard:
- EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS
- EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS
- EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS
- EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS
- EN ISO 374-1:2016 Type B (KPT), EN ISO 374-5:2016 VIRUS
- Godkända för livsmedelskontakt:
- +
- +
- +
- +
- +