Orders LinkContactUsLinkComponent
Complement reagents
Complement reagents
Complement reagents
Certificates
Complement reagents
1 Options Available Below
Product Details & Documents
Complement reagents are prepared from healthy, non-medicated animals using very large pools to reduce lot-to-lot variability. The fresh frozen complement is rapidly processed and maintained at –70 °C to ensure maximum activity. The lyophilised complement is supplied with a diluent of veronal buffered saline containing optimal concentrations of magnesium and calcium.
Specifications for Product Family
Specifications
  • Catalog No:
  • ROCKC300-0050
  • ROCKC300-0100
  • ROCKC300-0500
  • ROCKC301-0010
  • ROCKC301-0050
  • ROCKC301-0100
  • ROCKC302-0010
  • ROCKC303-0010
  • ROCKC303-0050
  • ROCKC304-0010
  • ROCKC304-0050
  • ROCKC304-0100
  • ROCKC304-0500
  • ROCKC305-0010
  • ROCKC305-0050
  • ROCKC305-0100
  • ROCKC306-0010
  • ROCKC306-0050
  • ROCKC306-0100
  • ROCKC309-0050
  • ROCKC309-0100
  • ROCKC200-0005
  • ROCKC201-0005
  • ROCKC202-0005
  • ROCKC203-0005
  • ROCKC204-0005
  • ROCKC205-0005
  • ROCKC206-0005
  • ROCKC209-0005
  • ROCKC220-0002
  • Formulering:
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Fresh frozen
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
  • Lyophilized
Specifications
Product Family Options
Lyophilised complement
Product Information
  • Beskrivning
    Availability
    Price
  • Guinea pig complement
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    ROCKC200-0005
    Formulering
    Lyophilized
  • Mouse complement
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    ROCKC201-0005
    Formulering
    Lyophilized
  • Rabbit A (nesting) complement
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    ROCKC202-0005
    Formulering
    Lyophilized
  • Rabbit B' (450 - 900 g) complement
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    ROCKC203-0005
    Formulering
    Lyophilized
  • Rabbit C (2265 - 2720 g) complement
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    ROCKC204-0005
    Formulering
    Lyophilized
  • Rat complement
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    ROCKC205-0005
    Formulering
    Lyophilized
  • Hamster complement
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    ROCKC206-0005
    Formulering
    Lyophilized
  • Goat complememnt
    Discontinued
    Specifications:
    More
    Specifications
    Cat. No.
    ROCKC209-0005
    Formulering
    Lyophilized
  • Rabbit anti-Sheep cell hemolysin
    Log in for availability
    Log in for availability
    1 * 2 mL
    1 * 2 mL - 5 075,00 kr
    5 075,00 kr
    1 * 2 mL
       5 075,00 kr
    Specifications:
    More
    Specifications
    Cat. No.
    ROCKC220-0002
    Formulering
    Lyophilized