DNA ladders
Catalog # 31084.
Supplier: Biotium
New Product
CAS Number:
Specifications
- Förp.:1
- Beskrivning:DNA ladder, 1 kb
- Seq:02
Specifications
About this item
Ready to use DNA ladders are supplied in 1X loading buffer at an optimal concentration for loading GelRed™ and GelGreen™ precast gels.