Bursa pastoris prepared slide
LIEDAS619D
- Beskrivning:Shepherd’s purse, capsella bursa pastoris
- Förp.:1
Prepared slide with longitudinal section of ovule with embryos in situ for general study.
Prepared slide with longitudinal section of ovule with embryos in situ for general study.