Phosphate buffered saline (PBS), 10X
Catalog # 786-027
Supplier: G-Biosciences
CAS Number:
Specifications
- Förp.:500 mL
- Beskrivning:PBS, 10×
Specifications
About this item
PBS 10X solution is used as a wash buffers for Western blotting, ELISA and other applications.