Apoptosis inducer set
Katalog # API-16S
Leverantör: G-Biosciences
CAS-nummer:
Specifikationer
- Förp.:1 SET
- Beskrivning:Apoptotic Inducer Set (Set of 6)
- Seq:06
Specifikationer
Om denna produkt
Apoptosis inducers are used for induction of apoptosis in cultured cells. The selection of apoptosis inducers include actinomycin D, camptothecin, cycloheximide, dexamethasone, doxorubicin and etoposide.
- Ready to use
- Long storage life