Capillary tubes
100 mm, 1 mm, 0,58 mm
Katalog # HILG1406119
Leverantör: HILGENBERG
Gå till:
Product Details & Documents
Borosilicate glass, ends cut, with filament.
Specifikationer
- Ø utv.:1 mm
- Ø inr.:0,58 mm
- Längd:100 mm
- Seq:08
Borosilicate glass, ends cut, with filament.