ContactUsLinkComponent
Double bossheads
  241-0156
Double bossheads
  241-0156
  • Material:
    Legering/plast
  • Förp.:
    1
  • För stavdiameter:
    16 mm
Double bosshead made from cast alloy.
  • With blue plastic headed nickel plated screws
  • For use with rods up to 16 mm