Analytical guard column, Upchurch Scientific®
Katalog # UPCHC-130B
Leverantör: UPCHURCH SCIENTIFIC
Specifikationer
- Kolumnformat:Guard column
- Ø inr.:2,00 mm
- Beskrivning:C-130B guard column including 2×A-100
- Längd:20 mm
- Seq:01
- Förp.:1
Specifikationer
Om denna produkt
This narrow-bore short column (2,0×20 mm) creates only a slight pressure increase with virtually no detectable theoretical plate loss when used with a 3 mm int.Ø or larger column.
- Available as a kit with funnel and extra frits
- Easy to pack