Wafer casettes
Katalog # PALBPCPC0101
Leverantör: Palbam
Specifikationer
- Beskrivning:4 inch wafer cassette
- Material:Rostfritt stål
- Seq:01
- Förp.:1
Specifikationer
Om denna produkt
The palbam class stainless steel wafer cassette is designed to replace standard plastic cassettes, in the semiconductor manufacturing process. The cassette has been designed with H Bar.
- Non-contaminating
- Non-deforming
- Fully conductive
- Withstands temperatures up to 400 ºC
- Designed for both manual and robotic handling