Nucleic acid and protein destain, Silver D-Stain™
Catalog # 786-199
Supplier: G-Biosciences
CAS Number:
Specifications
- Förp.:25
- Beskrivning:Nucleic acid and protein destains, Silver D-Stain™
Specifications
About this item
The staining of gels with silver stains often results in over-staining or unevenly stained gels. Silver D-Stain™ eliminates the need to repeat the electrophoresis.
Suitable for reducing the staining of silver stained gels and revealing over-stained bands.