Filter cards, brown, for TPX chambers, SHANDON™
Katalog # SHAN5991023
Leverantör: Epredia
Specifikationer
- Beskrivning:Filter cards, brown
- Förp.:200
Specifikationer
Om denna produkt
Highly absorbent disposable filter cards for use with TPX chamber.
- Use for sample volumes of 0,4 ml or less
- For single chambers
Not for use with Cytofunnel™ disposable sample chambers.