PCR microtubes
PCR tubes, individual, with attached cap, Klar, 0,5 ml
Leverantör: EPPENDORF
Gå till:
Product Details & Documents
PP.
- Etched lid for labelling
- Tight sealing
- Easy to open
- Certified free from human DNA, DNase, RNase and PCR inhibitors
These thin-walled, 0,5 ml PCR tubes are designed for use with all thermal cyclers with 0,5 ml block format.
Colour: Translucent
Documents
Certificates
Sök efter LOT-nummerSpecifikationer
- Kapacitet:0,5 ml
- Färg:Klar
- Beskrivning:PCR tubes, individual, with attached cap
- Cat. No.:731-0407
- Miljöcertificering:Renewable Energy Certificate, ISO 14001
- Environmentally Preferable:Low Manufacturing Impact