Petri dish holder
Catalog # 391-1150
Supplier: SciLabware
Specifications
- Beskrivning:Coated petri dish holder for six 90 mm dishes
- Seq:01
- Förp.:1
Specifications
About this item
Polyamide epoxy coated steel. Holds up to six 90 mm petri dishes.
- Better heat and chemical resistance than other coatings
- Ideal for small batch incubation