Slide box
Catalog # LIEDPK25
Supplier: LIEDER JOHANNES
Specifications
- Antal objektglas:25
- Färg:Blå
- Beskrivning:Box for microscope preparations
- Material:Plast
- Förp.:1
Specifications
About this item
This slide box is designed to facilitate the transportation of 25 microscope slides.
- Transparent cover for easy viewing
- Robust design and stackable construction
- Serrated retainer strips to maintain slides properly