Clamp locks for attaching reaction vessel
Catalog # RLAA2124
Supplier: R L ENTERPRISES
Specifications
- För:Quickfit® reaction vessels
- Förp.:1
Specifications
About this item
Alloy/zinc plated steel construction.
- Extension rods are secured to the body of the clamp with a socket screw (an allen key is provided)
- Jaws are heat resistant
Actuated by nickel-plated pummels and twin screw and fitted with stainless steel pivot pins. Each clamp is provided with two extension rods, one for vessels up to 5 L and the other up to 20 L.