Scrub pads, ErgoSCRUB®
Catalog # FOAMFT900
Supplier: FOAMTEC
Specifications
- Beskrivning:ErgoSCRUB® pad, soft handle, disposable
- Förp.:10
Specifications
About this item
ErgoSCRUB® is a soft ergonomically correct scrubbing tool for easy scrubbing of implant tools, large vacuum chambers, ceramic domes, and other spherical surfaces.
- Disposable
- Easy to grip
- Flexible enough to clean spherical surfaces, such as domes
- Enables faster cleaning of large tool surfaces
Suitable for chamber cleaning, CVD and CMP applications and for wafer handling.