Autosampler vial sleeve
Catalog # PERCN9307027
Supplier: PerkinElmer
Specifications
- För:200 µl and 300 µl autosampler vials
- Beskrivning:12×32 mm clear glass support sleeve, open base
- Förp.:25
Specifications
About this item
This open base clear glass support sleeve is used to offer microsampling capability in a standard 2 ml vial carousel or robotic autosampler.
- 12×32 mm support sleeve
- Accepts 200 and 300 µl vials