Drawer cabinet
Katalog # LEIH3808587E
Supplier: LEICA HISTOLOGY
Specifications
- Färg:Blå
- Beskrivning:Slide cabinet with eight drawers
- Material:Stål
- Förp.:1
Specifications
About this item
This drawer cabinet includes eight drawers and can be stacked for comfortable use.
Suitable for storing larger slides, transparencies and cassette blocks.