Chef jacket, Cumbria, C733
Supplier: Portwest
Certificates
About this item
These modern jackets are made of Kingsmill fabric constructed from 65% polyester and 35% cotton. The fabric guarantees high performance and maximum comfort. It is fully pre-shrunk and has superior dye retention.
- Mandarin collar, ring stud front
- Handy sleeve pocket
- Short sleeves
Weight: 190 g/m²
Specifications
- Catalog No:
- PRTWC733BKRXS
- PRTWC733BKRXXL
- PRTWC733BKRS
- PRTWC733BKRM
- PRTWC733WHRXS
- PRTWC733WHRXXL
- PRTWC733WHRS
- PRTWC733WHRXL
- PRTWC733WHRM
- PRTWC733WHRL
- PRTWC733BKRL
- PRTWC733BKRXL
- Modell:
- Cumbria, C733
- Cumbria, C733
- Cumbria, C733
- Cumbria, C733
- Cumbria, C733
- Cumbria, C733
- Cumbria, C733
- Cumbria, C733
- Cumbria, C733
- Cumbria, C733
- Cumbria, C733
- Cumbria, C733
- Outfit applikationer:
- Food
- Food
- Food
- Food
- Food
- Food
- Food
- Food
- Food
- Food
- Food
- Food
- Kön:
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Unisex
- Sleeve or leg length:
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Short
- Stängningsstorlek:
- Med knappar
- Med knappar
- Med knappar
- Med knappar
- Med knappar
- Med knappar
- Med knappar
- Med knappar
- Med knappar
- Med knappar
- Med knappar
- Med knappar
- Tyg:
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- Vikt:
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²
- 190 g/m²