Surface disinfectant, granulate, Dismozon® pur
Katalog # BODE975400
Leverantör: BODE CHEMIE
Specifikationer
- Beskrivning:Dismozon® pur, surface disinfectant
- Seq:01
- Förp.:30 g sachets
Specifikationer
Om denna produkt
Dismozon® pur is used for high-performance disinfection of washable surfaces and equipment in areas that are relevant to hygiene and sensitive to odours. Especially for the disinfection of highly sensitive materials. For use in outbreak situations, e.g. occurrence of infections associated with noroviruses or Clostridium difficile.
- Effective against Clostridium difficile spores
- Exceptional range of material compatibility
- Contains magnesium monoperoxyphthalate hexahydrate 800 mg/g
- Bactericidal, fungicidal, tuberculocidal, sporicidal and virucidal against enveloped viruses
- pH value: approx 5,3
Caution: Use biocides safely. Always read the label and product information before use.