Sling psychrometer
Katalog # SIMETM0081
Leverantör: Industrial Physics
Specifikationer
- Beskrivning:Sling psychrometer
- Förp.:1
Specifikationer
Om denna produkt
The psychrometer is designed to measure the percent relative humidity on the basis of the wet bulb-dry bulb thermometer principle.
- Conversion scale
- Plastic case on the instrument
- Relative humidity percent calculations