Sharps disposal boxes, Multi-Safe quick
Catalog # SEMA4779
Supplier: SEMADENI
Specifications
- Diam×H:100x125 mm
- Kapacitet:500 ml
- Beskrivning:Multi-Safe quick 500
- Seq:01
- Förp.:1
Specifications
About this item
The practical safety box for user-friendly disposal of sharp, pointed and dangerous objects for both stationary and ambulatory use.
- Round container form
- Openings of different sizes
- Containers can be fastened to a number of different holders to prevent them from overturning
- Closure cap does not spring back. This means nothing gets in the way during removal and stripping
- Various detaching devices