Orders LinkContactUsLinkComponent
Silicone bench protector
Silicone bench protector
500×500 mm, 2 mm
Katalog # 115-9137
Leverantör:   DELTALAB
Silicone bench protector
Katalog # 115-9137
Leverantör:  DELTALAB
Silicone bench protector
Silicone bench protector
500×500 mm, 2 mm
Price
1 775,00 kr
Log in för att kolla tillgänglighet och ert pris
1 * 1 Items
Produktdetaljer och dokument
Silicone sheet designed for laboratory benchtop protection or for use in autoclaves to protect any material.
  • Easy to clean with a wet cloth
  • Heat resistant up to 260 ºC
Specifikationer
  • L×B:
    500×500 mm
  • Tjocklek:
    2 mm