E-VAC aspiration system
Leverantör: BioTool
Certifikat
Om denna produkt
These compact waste systems are ideal for use with critical or dangerous liquids or supernatants, pathogens, or any class III or IV biohazard laboratories.
- Quiet, oil-free membrane pump (range from –250 to 650 mm Hg)
- Easy to clean stainless steel housing is UV resistant for use with laminar flows
- The E-Vac bottles can be autoclaved for 20 minutes at 121 °C to prevent laboratory contamination
- Base units are illuminated by LED to allow visual inspection of waste levels in the bottle
- Dual hydrophobic filters prevent liquids from reaching the pump housing and contaminating the system
Once target vacuum is reached, pump automatically switches off. Pump automatically switches on when vacuum pressure is applied, allowing it to maintain a constant vacuum pressure - perfect for gentle aspiration of small plates and dishes, as well as rapid emptying of larger containers.
E-Vac is available with a 4 litre PP bottle and lid with either barbed or with quick-release connectors for safe and convenient tubing connections and level detection sensor that automatically shuts the pump off when the bottle is full.
Delivery: E-Vac is available with a 4 litre PP bottle and lid with either barbed or with quick-release connectors for safe and convenient tubing connections and level detection sensor that automatically shuts the pump off when the bottle is full.