Urine collection system, UriSwab
Katalog # SLIN800C
Leverantör: STERILIN
Specifikationer
- Beskrivning:Urine collection systems
- Förp.:500
Specifikationer
Om denna produkt
A unique sponge swab is impregnated with the exact amount of boric acid for the sample collected and hence no fill line is required.
- Ideal for use in automation
- Simplifies collection of urine samples
Suitable for the collection, transport, and preservation of urine samples.
Caution: *Produkten/produkterna på denna sida ligger utanför tillämpningsområdet för EU -direktivet 2019/904