ContactUsLinkComponent
Chemiluminescence imaging systems, Syngene GeneGnome XRQ
Chemiluminescence imaging systems, Syngene GeneGnome XRQ
Catalog # SYNOGGNOME-XRQ-NPC
undefined
Chemiluminescence imaging systems, Syngene GeneGnome XRQ
Catalog # SYNOGGNOME-XRQ-NPC
Specifications
  • Beskrivning:
    GeneGnome XRQ, 4 MP, 16 bit system with f0.95 lens, without processor, without monitor
  • Seq:
    01
  • Förp.:
    1
Specifications
About this item
The Syngene GeneGnome XRQ is dedicated to chemiluminescence imaging. The system is built for high performance and automation and features a next generation high quantum efficiency CCD camera for even greater sensitivity. A very simple set-up process means a single click will automatically capture a quality image of any Western blot.
  • Configured for maximum sensitivity to ensure even the faintest band on a blot can be captured
  • High quantum efficiency cooled CCD camera (73% @ 425 nm) is very sensitive to low level light emissions from a blot
  • Peltier cooled camera has exceptional signal to noise performance resulting in virtually undetectable background noise
  • Small footprint - takes up minimal bench space
Delivery: Each system is supplied complete with unlimited copies of GeneTools analysis software.