Face shield, medical, 710
Katalog # UNVT710.00.00.00
Leverantör: UNIVET
Specifikationer
- Typ:Anti-fog
- Beskrivning:Face shield with clear lens and blue frame
- Förp.:1
Specifikationer
Om denna produkt
This face shield features an adjustable elastic band and an anallergic, interchangeable foam at the front.
- Interchangeable
- 90° flip up
Weight: 97 g
EN 166 (3 S CE)